Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120851.2 | NP_196386.1 | 2 - 3 | 682 | 138 | |
GenBank mRNA | AY035145.1 | - | 2 - 3 | 587 | 138 | |
AY059078.1 | - | 2 - 3 | 261 | 138 | ||
AY519617.1 | - | 2 - 3 | 261 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |