Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123521.4 | NP_198972.1 | 1 - 2 | 416 | 161 | |
GenBank mRNA | AF404864.1 | - | 1 - 2 | 353 | 161 | |
AK227522.1 | - | 1 - 2 | 359 | 161 | ||
BT004595.1 | - | 1 - 2 | 353 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |