Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125305.5 | NP_200723.1 | 2 - 3 | 452 | 116 | |
GenBank mRNA | AF370127.1 | - | 2 - 3 | 327 | 116 | |
AY040051.1 | - | 1 - 2 | 171 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |