Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345485.1 | NP_001331838.1 | 1 - 2 | 481 | 134 | |
NM_125558.3 | NP_568937.1 | 1 - 2 | 433 | 134 | ||
GenBank mRNA | AK227434.1 | - | 1 - 2 | 406 | 134 | |
BT005850.1 | - | 1 - 2 | 354 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |