Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126492.4 | NP_178540.1 | 5 - 6 | 503 | 153 | |
GenBank mRNA | AK229481.1 | - | 5 - 6 | 435 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |