Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084556.2 | NP_001078025.1 | 3 - 4 | 669 | 125 | |
NM_001336805.1 | NP_001324466.1 | 1 - 2 | 374 | 125 | ||
NM_129547.3 | NP_181518.1 | 2 - 3 | 774 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |