Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084556.2 | NP_001078025.1 | 7 - 8 | 1290 | 129 | |
NM_001336805.1 | NP_001324466.1 | 5 - 6 | 995 | 129 | ||
NM_129547.3 | NP_181518.1 | 6 - 7 | 1395 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |