Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100131.3 | NP_171751.1 | 2 | 1620 | 98 | |
NM_179246.1 | NP_849577.1 | 2 | 1460 | 98 | ||
GenBank mRNA | AF325061.2 | - | 2 | 1452 | 98 | |
AY087703.1 | - | 2 | 1460 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |