Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QQS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339054.1 | NP_001326493.1 | 3 | 425 | 74 | |
NM_001339055.1 | NP_001326492.1 | 4 | 347 | 74 | ||
NM_113975.5 | NP_189695.1 | 3 | 428 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |