Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148857.3 | NP_680110.1 | 1 | 958 | 121 | |
GenBank mRNA | AY096499.1 | - | 1 | 853 | 121 | |
AY133854.1 | - | 1 | 762 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |