Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179699.3 | NP_850030.1 | 2 | 2120 | 80 | |
NM_201785.1 | NP_973514.1 | 2 | 2063 | 80 | ||
GenBank mRNA | BT002751.1 | - | 1 | 1938 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |