Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DREB2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111939.3 | NP_187713.1 | 2 | 1136 | 66 | |
GenBank mRNA | AB007791.1 | - | 2 | 1090 | 66 | |
AK117363.1 | - | 2 | 1136 | 66 | ||
BT008351.1 | - | 1 | 908 | 66 | ||
KC412253.1 | - | 1 | 1562 | 66 |
Gene Symbol: TBL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111940.3 | NP_187714.1 | 5 | 1820 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |