Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338122.1 | NP_001319550.1 | 4 | 2329 | 164 | |
NM_001338123.1 | NP_001326728.1 | 3 | 2426 | 164 | ||
NM_112315.4 | NP_188073.2 | 2 | 1227 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |