Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338603.1 | NP_001326806.1 | 2 | 537 | 88 | |
NM_113190.3 | NP_566719.1 | 2 | 639 | 88 | ||
GenBank mRNA | AY085637.1 | - | 2 | 527 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |