Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCR73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035875.2 | NP_001030952.1 | 1 - 2 | 136 | 82 | |
GenBank mRNA | AY803261.1 | - | 1 - 2 | 68 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |