Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036555.3 | NP_001031632.1 | 3 - 4 | 247 | 126 | |
NM_001203797.1 | NP_001190726.1 | 4 - 5 | 280 | 126 | ||
NM_001340903.1 | NP_001319932.1 | 4 - 5 | 465 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |