Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037010.2 | NP_001032087.1 | 1 | 1778 | 128 | |
NM_001345246.1 | NP_001332283.1 | 1 | 1778 | 128 | ||
GenBank mRNA | AK221521.1 | - | 1 | 1619 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |