Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341648.1 | NP_001320053.1 | 1 - 2 | 222 | 122 | |
NM_001341650.1 | NP_001328601.1 | 1 - 2 | 222 | 122 | ||
NM_118551.3 | NP_194149.2 | 1 - 2 | 222 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |