Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036165.1 | NP_001031242.1 | 2 - 3 | 1371 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |