Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341766.1 | NP_001329919.1 | 5 - 6 | 886 | 128 | |
NM_118719.4 | NP_194316.2 | 5 - 6 | 873 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |