Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123430.2 | NP_198881.1 | 1 - 2 | 736 | 147 | |
GenBank mRNA | AY074299.1 | - | 1 - 2 | 674 | 147 | |
AY123036.1 | - | 1 - 2 | 668 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |