Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127038.2 | NP_179081.1 | 6 - 7 | 1532 | 136 | |
NM_179624.2 | NP_849955.1 | 6 - 7 | 1532 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |