Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340846.1 | NP_001328007.1 | 6 - 7 | 1370 | 124 | |
NM_117401.2 | NP_193064.2 | 7 - 8 | 1280 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |