Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120395.2 | NP_195937.1 | 1 | 650 | 82 | |
GenBank mRNA | AF360183.1 | - | 1 | 650 | 82 | |
AY039999.1 | - | 1 | 631 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |