Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084021.2 | NP_001077490.1 | 1 - 2 | 286 | 68 | |
GenBank mRNA | AF136539.1 | - | 1 - 2 | 92 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |