Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084021.2 | NP_001077490.1 | 3 - 4 | 548 | 65 | |
GenBank mRNA | AF136539.1 | - | 3 - 4 | 354 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |