Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084268.2 | NP_001077737.1 | 1 - 2 | 44 | 84 | |
GenBank mRNA | DQ446369.1 | - | 1 - 2 | 44 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |