Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118533.4 | NP_194132.3 | 2 - 3 | 1008 | 151 | |
GenBank mRNA | AK220630.1 | - | 2 - 3 | 780 | 151 | |
KJ138738.1 | - | 2 - 3 | 734 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |