Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173990.2 | NP_776415.1 | 7 - 8 | 1019 | 128 | |
GenBank mRNA | BC123795.1 | - | 7 - 8 | 1019 | 128 | |
M95605.1 | - | 7 - 8 | 1018 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |