Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034726.2 | NP_001029898.1 | 2 - 3 | 231 | 106 | |
GenBank mRNA | BC102910.1 | - | 2 - 3 | 232 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |