Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076973.1 | NP_001070441.1 | 1 - 2 | 318 | 51 | |
GenBank mRNA | BC149326.1 | - | 1 - 2 | 314 | 51 | |
BT021776.1 | - | 1 - 2 | 318 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |