Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001139.2 | NP_001001139.1 | 4 - 5 | 786 | 71 | |
GenBank mRNA | X55010.1 | - | 787 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |