Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001163.1 | NP_001001163.1 | 1 - 2 | 113 | 96 | |
GenBank mRNA | AJ699421.1 | - | 1 - 2 | 113 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |