Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001164.1 | NP_001001164.1 | 4 - 5 | 384 | 106 | |
GenBank mRNA | AY581200.1 | - | 4 - 5 | 384 | 106 | |
BC102581.1 | - | 4 - 5 | 401 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |