Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPTIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001168.1 | NP_001001168.1 | 12 - 13 | 1162 | 114 | |
GenBank mRNA | AY568576.1 | - | 12 - 13 | 1162 | 114 | |
BC148893.1 | - | 12 - 13 | 1129 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |