Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001599.1 | NP_001001599.1 | 3 - 4 | 850 | 73 | |
GenBank mRNA | AJ704563.1 | - | 3 - 4 | 850 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |