Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001600.2 | NP_001001600.2 | 5 - 6 | 685 | 112 | |
GenBank mRNA | AY330769.1 | - | 4 - 5 | 606 | 112 | |
BC149644.1 | - | 4 - 5 | 635 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |