Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003902.1 | NP_001003902.1 | 1 - 2 | 133 | 81 | |
GenBank mRNA | AY675077.1 | - | 1 - 2 | 133 | 81 | |
BC108164.1 | - | 1 - 2 | 59 | 81 | ||
BC142031.1 | - | 1 - 2 | 63 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |