Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007806.2 | NP_001007807.2 | 3 - 4 | 383 | 141 | |
GenBank mRNA | AY692440.1 | - | 2 - 3 | 187 | 141 | |
BC103350.1 | - | 3 - 4 | 383 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |