Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007811.2 | NP_001007812.2 | 11 - 12 | 1308 | 77 | |
GenBank mRNA | AY788896.1 | - | 2 - 3 | 228 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |