Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008412.2 | NP_001008412.1 | 2 - 3 | 286 | 74 | |
GenBank mRNA | AF412040.1 | - | 2 - 3 | 287 | 74 | |
AF416586.1 | - | 1 - 2 | 193 | 74 | ||
EU682381.1 | - | 2 - 3 | 229 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |