Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011685.1 | NP_001011685.1 | 8 - 9 | 1350 | 57 | |
GenBank mRNA | AY854631.1 | - | 8 - 9 | 1350 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |