Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014955.1 | NP_001014955.1 | 1 - 2 | 843 | 106 | |
GenBank mRNA | BC102299.1 | - | 1 - 2 | 728 | 106 | |
BT020640.1 | - | 1 - 2 | 843 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |