Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5MG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024550.4 | NP_001019721.4 | 2 - 3 | 268 | 104 | |
GenBank mRNA | BC102455.1 | - | 2 - 3 | 270 | 104 | |
S70448.1 | - | 2 - 3 | 239 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |