Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032299.1 | NP_001027470.1 | 2 - 3 | 388 | 108 | |
GenBank mRNA | AJ699418.1 | - | 2 - 3 | 388 | 108 | |
BC149573.1 | - | 2 - 3 | 852 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |