Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FETUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034218.2 | NP_001029390.1 | 2 - 3 | 440 | 129 | |
GenBank mRNA | BC103069.1 | - | 2 - 3 | 441 | 129 | |
BT021735.1 | - | 2 - 3 | 419 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |