Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FETUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034218.2 | NP_001029390.1 | 6 - 7 | 894 | 88 | |
GenBank mRNA | BC103069.1 | - | 6 - 7 | 895 | 88 | |
BT021735.1 | - | 6 - 7 | 873 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |