Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034429.3 | NP_001029601.1 | 2 - 3 | 289 | 69 | |
NR_130174.1 | 2 - 3 | 526 | 69 | |||
NR_130175.1 | 3 - 4 | 341 | 69 | |||
GenBank mRNA | BC102969.1 | - | 2 - 3 | 272 | 69 | |
DV782931.1 | - | 2 - 3 | 263 | 69 | ||
DV800190.1 | - | 2 - 3 | 521 | 69 | ||
DV822848.1 | - | 2 - 3 | 283 | 69 | ||
DV845693.1 | - | 2 - 3 | 290 | 69 | ||
EH143926.1 | - | 3 - 4 | 295 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |