Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034755.1 | NP_001029927.1 | 3 - 4 | 767 | 116 | |
GenBank mRNA | BC149870.1 | - | 3 - 4 | 787 | 116 | |
BT020665.1 | - | 3 - 4 | 767 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |