Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034799.1 | NP_001029971.1 | 2 - 3 | 594 | 66 | |
GenBank mRNA | BC103144.1 | - | 2 - 3 | 594 | 66 | |
BT026259.1 | - | 2 - 3 | 474 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |